Clamps for universal platforms
10 ml kolbeklemme, 60 (280×330); 113 (457×457); 157 (457×609) for MaxQ 2000
112 (760×457) and 160 (910×609) clamps for MaxQ 3000
112 (760×457) and 160 (910×609) clamps for MaxQ 3000
Katalog # 444-0304
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Documents
BARN30156BI_TDS_EN
Datablad
Specifikationer
- Til:60 (280×330); 113 (457×457); 157 (457×609) for MaxQ 2000
112 (760×457) and 160 (910×609) clamps for MaxQ 3000 - Beskrivelse:10 ml kolbeklemme



