Orders LinkContactUsLinkComponent
Halvmasker, serie 7700
Halvmasker, serie 7700
770030M, Silicone, M
Katalog # 111-0290
Leverandør:   Honeywell Safety
Halvmasker, serie 7700
Katalog # 111-0290
Leverandør:  Honeywell Safety
Halvmasker, serie 7700
Halvmasker, serie 7700
770030M, Silicone, M
Price
523,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Soft, hypo-allergenic silicone half masks. Interchangeable cartridges and filters provide air-purifying protection for a variety of applications. Direct cartridge-to-face-piece connection eliminates gaskets and cartridge receptacles.
  • Soft silicone makes these respirators close fitting and comfortable
  • Fit snugly while the cradle suspension system alleviates pressure points
  • Can be worn with safety glasses and goggles
 : NIOSH approved for impact and penetration resistance when used with North Safety cartridges and/or filters.
 : Cartridges and filters must be ordered separately
Specifikationer
  • Model:
    770030M, Silicone
  • Seq:
    02
  • Størrelse:
    M
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Filter cartridge class P3, material: ABS - fiber glass paper, protection type: particulate
    Filter cartridge class P3, material: ABS - fiber glass paper, protection type: particulate
    NORHN06575008
    Log ind for tilgængelighed
    1 * 4 Items - kr981.00