Halvmasker, serie 7700
770030L, Silicone, L
Katalog # 111-0289
Leverandør: Honeywell Safety
Gå til:
Product Details & Documents
Soft, hypo-allergenic silicone half masks. Interchangeable cartridges and filters provide air-purifying protection for a variety of applications. Direct cartridge-to-face-piece connection eliminates gaskets and cartridge receptacles.
- Soft silicone makes these respirators close fitting and comfortable
- Fit snugly while the cradle suspension system alleviates pressure points
- Can be worn with safety glasses and goggles
Certifications: NIOSH approved for impact and penetration resistance when used with North Safety cartridges and/or filters.
Bestillingsoplysninger: Cartridges and filters must be ordered separately
Specifikationer
- Model:770030L, Silicone
- Seq:03
- Størrelse:L
Accessories
Produktinformation
- Katalog #TilgængelighedPrice
- Filter cartridge class P3, material: ABS - fiber glass paper, protection type: particulateFilter cartridge class P3, material: ABS - fiber glass paper, protection type: particulateNORHN06575008Log ind for tilgængelighed1 * 4 Items - 981,00 kr