Half area microplates, 96-well
Half area, solid bottom, non treated surface, klar, +
Leverandør: Greiner Bio-One
Half area microplates, 96-well
Katalog # GREI675161_40
Leverandør: Greiner Bio-One
Gå til:
Product Details & Documents
PS, without lid. 96-well half area microplates offer an alternative to 384-well or 1536-well microplates when low sample volume is an important criterion but automation is not necessarily appropriate. They can be pipetted manually while at the same time allow a reduction in sample volume of up to 50%.
Microplates with clear bottoms feature micro-thin µClear® bottoms for exceptional optical properties and standardised pathlength, eliminating the need for pathlength determinations. White plates are ideal for luminescence assays, while black plates are suitable for fluorescence assays. Both formats are also suitable for cell-based assays.
Specifikationer
- Steril:+
- Farve:klar
- Beskrivelse:Half area, solid bottom, non treated surface
- Seq:01
- Cat. No.:GREI675161_40
- Miljøcertificering:ISO 14001 (241002); ISO 50001 (241002)
- Environmentally Preferable:Lav fremstillingspåvirkning