Propyl 4-hydroxybenzoat ≥99%
Glass bottle for solids, 250 g
Purity: 99+% CAS No.: 94-13-3 Molecular Formula: C10H12O3 Formula Weight: 180.20 Flash Point: 180°(356°F) Melting Point: 95-99° Beilstein Reference: 1103245 Merck Reference: 14,7866 MDL No.: MFCD00002354
: Propyl 4-hydroxybenzoate, Propyl parahydroxybenzoate, Propylparaben, 4-Hydroxybenzoic acid propyl ester
- :94-13-3
- :C₁₀H₁₂O₃
- :MFCD00002354
- :180.2 g/mol
- :14,7866
- :292 °C (1013 hPa)
- :95…99 °C
- :1,28 g/cm³ (20 °C)
- :180 °C (356 °F)
- :Stue temperatur
- :202-307-7
- Pakning:Glass bottle for solids
- Pakn:250 g