Orders LinkContactUsLinkComponent
Propyl 4-hydroxybenzoat ≥99%
Glass bottle for solids, 250 g
  A10180.30
Propyl 4-hydroxybenzoat ≥99%
  A10180.30
 :  94-13-3
Propyl 4-hydroxybenzoat ≥99%
Propyl 4-hydroxybenzoat ≥99%
Glass bottle for solids, 250 g
 :  94-13-3
600,00 kr
1 * 250 g
Purity: 99+% CAS No.: 94-13-3 Molecular Formula: C10H12O3 Formula Weight: 180.20 Flash Point: 180°(356°F) Melting Point: 95-99° Beilstein Reference: 1103245 Merck Reference: 14,7866 MDL No.: MFCD00002354
 :  Propyl 4-hydroxybenzoate, Propyl parahydroxybenzoate, Propylparaben, 4-Hydroxybenzoic acid propyl ester
  •  :94-13-3
  •  :C₁₀H₁₂O₃
  •  :MFCD00002354
  •  :180.2 g/mol
  •  :14,7866
  •  :292 °C (1013 hPa)
  •  :95…99 °C
  •  :1,28 g/cm³ (20 °C)
  •  :180 °C (356 °F)
  •  :Stue temperatur
  •  :202-307-7
  • Pakning:
    Glass bottle for solids
  • Pakn:
    250 g