Agarose, NuSieve® 3:1
Agarose, NuSieve® 3:1
Katalog # 733-1209
Leverandør: LONZA
Gå til:
Product Details & Documents
NuSieve® 3:1 is a high strength agarose ideal for analytical electrophoresis of small DNA/RNA fragments and in-gel PCR/transformation/ligation.
- High gel strength makes gels easy to handle, enhancing the convenience of gel processing and blotting
- Performance tested to ensure fine resolution of DNA fragments up to 1000 bp
- DNAse- and RNAse-free
Documents
Certifikater
Søg efter partinummerSpecifikationer
- Beskrivelse:Agarose, NuSieve® 3:1
- Seq:03