Orders LinkContactUsLinkComponent
Agarose, NuSieve® 3:1
Agarose, NuSieve® 3:1
Agarose, NuSieve® 3:1
Katalog # 733-1209
Leverandør:   LONZA
Agarose, NuSieve® 3:1
Katalog # 733-1209
Leverandør:  LONZA
Agarose, NuSieve® 3:1
Agarose, NuSieve® 3:1
Agarose, NuSieve® 3:1
Price
30.870,00 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents
NuSieve® 3:1 is a high strength agarose ideal for analytical electrophoresis of small DNA/RNA fragments and in-gel PCR/transformation/ligation.
  • High gel strength makes gels easy to handle, enhancing the convenience of gel processing and blotting
  • Performance tested to ensure fine resolution of DNA fragments up to 1000 bp
  • DNAse- and RNAse-free
Documents
Specifikationer
  • Beskrivelse:
    Agarose, NuSieve® 3:1
  • Seq:
    03