Tube racks, universal
ufarvet, 32/32/12/4, 0,5/1,5-2,0/15/50 ml, PP, 172×50×90 mm
Supplier: DELTALAB
Jump to:
Product Details & Documents
PP, linkable to other racks.
- Versatile rack with different capacities on each of the four sides
- Alpha-numeric matrix on the four sides for better sample location
- Suitable for freezers and autoclaves
Documents
Certificates
Search by Lot NumberSpecifications
- Til rør:0,5/1,5-2,0/15/50 ml
- Ant. huller:32/32/12/4
- Farve:ufarvet
- Materiale:PP
- Seq:01
- B×D×H:172×50×90 mm