Orders LinkContactUsLinkComponent
Tube racks, universal
Tube racks, universal
ufarvet, 32/32/12/4, 0,5/1,5-2,0/15/50 ml, PP, 172×50×90 mm
Catalog # DELTM-520
Supplier:   DELTALAB
Tube racks, universal
Catalog # DELTM-520
Supplier:  DELTALAB
Tube racks, universal
Tube racks, universal
ufarvet, 32/32/12/4, 0,5/1,5-2,0/15/50 ml, PP, 172×50×90 mm
Price
1.020,00 kr
Log in for availability
1 * 5 Items
Product Details & Documents
PP, linkable to other racks.
  • Versatile rack with different capacities on each of the four sides
  • Alpha-numeric matrix on the four sides for better sample location
  • Suitable for freezers and autoclaves
Documents
Specifications
  • Til rør:
    0,5/1,5-2,0/15/50 ml
  • Ant. huller:
    32/32/12/4
  • Farve:
    ufarvet
  • Materiale:
    PP
  • Seq:
    01
  • B×D×H:
    172×50×90 mm