Orders LinkContactUsLinkComponent
Cavity slides
Cavity slides
Cavity slides, 1
Catalog # 630-1611
Supplier:   MARIENFELD
Cavity slides
Catalog # 630-1611
Supplier:  MARIENFELD
Cavity slides
Cavity slides
Cavity slides, 1
Price
940,00 kr
Log in for availability
1 * 50 Items
Product Details & Documents
Soda-lime glass of hydrolytic class 3 cavity slides.
Carefully ground and polished for in vitro diagnostic (IVD) applications.
Certifications: IVD 98/79/EC
Documents
Specifications
  • Ant. brønde:
    1
  • Beskrivelse:
    Cavity slides
  • Seq:
    02