Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
1-Naphtol ≥99%

1-Naphtol ≥99%

Glass bottle for solids, 100 g
Catalog # A11862.22
Corrosive
Irritant
1-Naphtol ≥99%
Catalog # A11862.22
CAS Number:  90-15-3
1-Naphtol ≥99%
1-Naphtol ≥99%

Glass bottle for solids, 100 g

CAS Number:  90-15-3
Price
293,00 kr
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 99% CAS No.: 90-15-3 Molecular Formula: C10H8O Formula Weight: 144.17 Flash Point: 125°(257°F) Odour: schwach, phenolartig Boiling Point: 278-280° Solubility: Soluble in alcohol, benzene, chloroform, ether Beilstein Reference: 1817321 Merck Reference: 14,6383 UN No.: 2811 MDL No.: MFCD00003930

Synonyms:  α-Naphtol
  • CAS Number:90-15-3
  • Formula:C₁₀H₈O
  • MDL Number:MFCD00003930
  • Molecular Weight:144.17 g/mol
  • Merck Index:14,6383
  • Boiling Pt:278…280 °C
  • Melting Pt:95…97 °C
  • Density:1,28
  • Flash Pt:125 °C (257 °F)
  • Storage Temperature:Stue temperatur
  • EINECS:201-969-4
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g

Specifications