Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Super Adjustable™ Super Erecta® shelving

Super Adjustable™ Super Erecta® shelving

Super Adjustable™ wire shelf, A2460NK3, Metrosæl 3, 1524×610×1370 mm, 9,5 kg
Catalog # METOA2460NK3
Supplier:   Metro International
Super Adjustable™ Super Erecta® shelving
Catalog # METOA2460NK3
Super Adjustable™ Super Erecta® shelving
Super Adjustable™ Super Erecta® shelving

Super Adjustable™ wire shelf, A2460NK3, Metrosæl 3, 1524×610×1370 mm, 9,5 kg

Price
2.270,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Designed to work in conjunction with the entire Super Erecta® System of shelves and accessories, these Super Adjustable wire shelves have a unique corner release system which allows them to be adjusted quickly and easily without the use of tools.

  • Heavy duty construction
  • Built to save space and to stand though against years of heavy use
  • Adjust and add shelves quickly and easily to reclaimwasted space

They shelf have a sleek chrome finish that makes it perfect for dry storage areas, while the open-wire construction minimises dust accumulation and allows a free circulation of air and greater visibility.

Specifications
  • Beskrivelse:
    Super Adjustable™ wire shelf, A2460NK3
  • Materiale:
    Metrosæl 3
  • Seq:
    30
  • B×D×H:
    1524×610×1370 mm
  • Vægt:
    9,5 kg