Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Carbon standards

Organic carbon standard, 1000 ppm, 500 mL
  REGETOC1M
 :   REAGECON
Carbon standards
  REGETOC1M
 :  REAGECON
 :  
Carbon standards

Organic carbon standard, 1000 ppm, 500 mL

 :  
 
2.070,00 kr
   
1 * 500 mL
This product has been blocked by your organization. Please contact your purchasing department for more information
 

These standards are manufactured from high purity raw materials and is suitable for calibrating a wide range of Total Organic Carbon (TOC) and Total Inorganic Carbon (TIC) analysers to a high accuracy.

These standards are independent, traceable and certified.

Certifications: Accredited to IEC/ISO 17025 and ISO 17034.

 

 

 

  • Koncentration:
    1000 ppm
  • Pakn:
    500 mL
  • Beskrivelse:
    Organic carbon standard