Orders LinkContactUsLinkComponent
Colony counters
Kolonitæller, manuel, med forstørrelsesglas Ø 100 mm, uden lys, med fod
  710-0107
Colony counters
  710-0107
 :  
Colony counters
Colony counters
Kolonitæller, manuel, med forstørrelsesglas Ø 100 mm, uden lys, med fod
Choice of models with a fibre marker with a built-in switch and /or a metal-tipped pen that are attached to the base. The marker supplied with the counter allows marked cells to be added to the screen one by one.
  • Ø 100 or 120 mm biconvex lamp
  • X2 magnification
220 V voltage, 50 Hz, 0,2 A.
  • Beskrivelse:
    Kolonitæller, manuel, med forstørrelsesglas Ø 100 mm, uden lys, med fod
  • Seq:
    01