Orders LinkContactUsLinkComponent
Adhesive mounting tapes
Adhesive mounting tapes
Glass silk adhesive tape, max. 200 °C, 15 mm
Katalog # ISOP610210-000
Leverandør:   ISOPAD
Adhesive mounting tapes
Katalog # ISOP610210-000
Leverandør:  ISOPAD
Adhesive mounting tapes
Glass silk adhesive tape, max. 200 °C, 15 mm
Price
282,73 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Glass-silk adhesive tapes used for mounting.
Specifikationer
  • Bredde:
    15 mm
  • Beskrivelse:
    Glass silk adhesive tape, max. 200 °C
  • Seq:
    01