Orders LinkContactUsLinkComponent
Rods, AR-Glas®
Rods, AR-Glas®
4±0,15 mm, 1500 mm
Katalog # SCOR1028126
Leverandør:   Schott AG
Rods, AR-Glas®
Katalog # SCOR1028126
Leverandør:  Schott AG
Rods, AR-Glas®
4±0,15 mm, 1500 mm
Price
1.460,00 kr
Log ind for tilgængelighed
1 * 14,5 kg
Product Details & Documents
Made of AR®-Glas.
  • Soft glass is acid-resistant
  • Has high thermal expansion for pressure melting and for fusing with platinum or dumet and other alloys
Specifikationer
  • Udv.Ø:
    4±0,15 mm
  • Længde:
    1500 mm
  • Seq:
    02