Accessories for Assortment Boxes, PS
Compartments, PS for storage boxes, grey, 322×108 mm, 45 mm height
Katalog # SEMA0440
Leverandør: SEMADENI
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:Compartments, PS for storage boxes, grey, 322×108 mm, 45 mm height
- Seq:11
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



