Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Accessories for Assortment Boxes, PS

Accessories for Assortment Boxes, PS

Compartments, PS for storage boxes, grey, 215×108 mm, 45 mm height
Katalog # SEMA0439
Leverandør:   SEMADENI
Accessories for Assortment Boxes, PS
Katalog # SEMA0439
Leverandør:  SEMADENI
Accessories for Assortment Boxes, PS
Accessories for Assortment Boxes, PS

Compartments, PS for storage boxes, grey, 215×108 mm, 45 mm height

Price
83,90 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Compartments, PS for storage boxes, grey, 215×108 mm, 45 mm height
  • Seq:
    10