Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

(±)-Epichlorhydrin ≥99%

Glass bottle for solids, 500 g
Katalog # A15823.36
Giftig
Sundhed
Ætsende
Brandfarlig
Irriterende
Leverandør:   Thermo Fisher Scientific
(±)-Epichlorhydrin ≥99%
Katalog # A15823.36
CAS nummer:  106-89-8
(±)-Epichlorhydrin ≥99%
(±)-Epichlorhydrin ≥99%

Glass bottle for solids, 500 g

CAS nummer:  106-89-8
Price
448,00 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents

Purity: 99% CAS No.: 106-89-8 Molecular Formula: C3H5ClO Formula Weight: 92.53 Flash Point: 32°(90°F) Physical Form: Liquid Density: 1.182 Refractive Index: 1.4380 Appearance: Colourless Odour: Like chlorine Melting Point: -57° Boiling Point: 115-117° Beilstein Reference: 79785 Merck Reference: 14,3611 UN No.: 2023 MDL No.: MFCD00005132

A cyclodextrin cross-linking reagent

Synonymer:  Epichlorhydrin, 1-Chlor-2,3-epoxypropan, (±)-1-Chlor-2,3-epoxypropan, 3-Chlor-1,2-epoxypropan, (±)-3-Chlor-1,2-epoxypropan
  • CAS nummer:106-89-8
  • Formel:C₃H₅ClO
  • MDL nummer:MFCD00005132
  • Molecular Weight:92.52 g/mol
  • Merck Index:14,3611
  • Kogende Pt:115…117 °C
  • Smeltende Pt:–57 °C
  • Tæthed:1,182
  • Flash Pt:32 °C (90 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:203-439-8
  • FN:2023
  • ADR:6.1,II
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    500 g

Specifikationer