Orders LinkContactUsLinkComponent
Instrument dishes
Instrument dishes
Låg til instrumentbakker 700 ml, PS krystalklar, 210×110×20 mm
Katalog # BURK4230-2071
Leverandør:   BURKLE
Instrument dishes
Katalog # BURK4230-2071
Leverandør:  BURKLE
Instrument dishes
Instrument dishes
Låg til instrumentbakker 700 ml, PS krystalklar, 210×110×20 mm
Price
138,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Trays for various uses. In laboratories, doctors’ surgeries, hospitals, pharmacies, the foodstuffs industry - wherever there is a need for clean storage of instruments, equipment, goods and precision parts.
  • Melamine, white
  • Tray without lid
Lid available as accessory.
Specifikationer
  • Beskrivelse:
    Låg til instrumentbakker 700 ml, PS krystalklar
  • B×D×H:
    210×110×20 mm