Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Chloramphenicol ≥99%

Chloramphenicol ≥99%

Plastic bottle for solids, 25 g
Katalog # B20841.14
Sundhed
Leverandør:   Thermo Fisher Scientific
Chloramphenicol ≥99%
Katalog # B20841.14
CAS nummer:  56-75-7
Chloramphenicol ≥99%
Chloramphenicol ≥99%

Plastic bottle for solids, 25 g

CAS nummer:  56-75-7
Price
545,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents

Purity: 99+% CAS No.: 56-75-7 Molecular Formula: C11H12Cl2N2O5 Formula Weight: 323.14 Physical Form: Crystalline powder Appearance: White to pale cream Melting Point: 149-152° Beilstein Reference: 2225532 Merck Reference: 14,2077 Optical Rotation: +19° (c=6 in ethanol) MDL No.: MFCD00078159

Inhibitor of translation on the 50S subunit at the peptidyltransferase step

Synonymer:  Chloromycetin®, Tega-Cetin, (R,R)-Chloramphenicol
  • CAS nummer:56-75-7
  • Formel:Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
  • MDL nummer:MFCD00078159
  • Molecular Weight:323.13 g/mol
  • Merck Index:14,2077
  • Kogende Pt:645 °C (1013 hPa)
  • Smeltende Pt:149…152 °C
  • Tæthed:1,49 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-287-4
Specifikationer

Specifikationer

  • Pakning:
    Plastic bottle for solids
  • Pakn:
    25 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.