Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Guanin ≥98%

Guanin ≥98%

Glass bottle for solids, 25 g
Katalog # A12024.14
Irriterende
Leverandør:   Thermo Fisher Scientific
Guanin ≥98%
Katalog # A12024.14
CAS nummer:  73-40-5
Guanin ≥98%
Guanin ≥98%

Glass bottle for solids, 25 g

CAS nummer:  73-40-5
Price
578,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 73-40-5 Molecular Formula: C5H5N5O Formula Weight: 151.13 Storage Temperature: 2°C to 8°C Melting Point: 300° Beilstein Reference: 9680 Merck Reference: 14,4564 MDL No.: MFCD00071533

Synonymer:  2-Amino-1H-purin-6(7H)-one
  • CAS nummer:73-40-5
  • Formel:C₅H₅N₅O
  • MDL nummer:MFCD00071533
  • Molecular Weight:151.13 g/mol
  • Merck Index:14,4564
  • Smeltende Pt:>300 °C
  • Tæthed:1,67 g/cm³ (20 °C)
  • Opbevaringstemperatur:Køleskab
  • EINECS:200-799-8
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.