Particle traps for use with PLOT columns
Particle trap for PoraPLOT columns, fused silica, Int.Ø×L: 0,32 mm × 2,5 m
Catalog # VARICP4016
Supplier: VARIAN
Jump to:
Product Details & Documents
Though highly stabilised, it is impossible to guarantee that no particles will dislodge from the column wall. When used in valve-switching applications, the use of a particle trap can prevent scarring of the rotors.
Specifications
- Beskrivelse:Particle trap for PoraPLOT columns, fused silica, Int.Ø×L: 0,32 mm × 2,5 m
- Seq:03