Particle traps for use with PLOT columns
Particle trap for PoraPLOT columns, UltiMetal, includes CP-UltiMetal connector, Int.Ø×L: 0,53 mm × 2,5 m
Katalog # VARICP4018
Leverandør: VARIAN
Gå til:
Product Details & Documents
Though highly stabilised, it is impossible to guarantee that no particles will dislodge from the column wall. When used in valve-switching applications, the use of a particle trap can prevent scarring of the rotors.
Specifikationer
- Beskrivelse:Particle trap for PoraPLOT columns, UltiMetal, includes CP-UltiMetal connector, Int.Ø×L: 0,53 mm × 2,5 m
- Seq:05