Orders LinkContactUsLinkComponent
Solid phase microextraction fibers
Solid phase microextraction fibers
SPME link septa, 11 mm
Katalog # VARI392548402
Solid phase microextraction fibers
Katalog # VARI392548402
Leverandør:  
Solid phase microextraction fibers
SPME link septa, 11 mm
Discontinued
Product Details & Documents
Solid phase microextraction (SPME) is a technique for extracting analytes from solid, liquid or gaseous samples by adsorbing them onto the SPME fiber and then desorbing them into an inlet, either on a gas chromatograph (GC) or an HPLC system. SPME is amenable to automation using an autosampler or it can be performed manually as well.
Solid phase microextraction kits: Kits are also available to support method development, offering a variety of fiber types and configurations within a single kit. SPME fiber kits contain three fibers.
Specifikationer
  • Beskrivelse:
    SPME link septa, 11 mm
  • Seq:
    37
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Merlin microseal, SPME replacement seal, 23 gauge
    Merlin microseal, SPME replacement seal, 23 gauge
    VARI392609902
    Log ind for tilgængelighed
    1 * 1 Items - 4.350,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • SPME fiber holder for CTC autosampler
    SPME fiber holder for CTC autosampler
    VARISU57347U
    Log ind for tilgængelighed
    1 * 1 Items
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • SPME stand, 15 ml
    SPME stand, 15 ml
    VARISU57357U
    Log ind for tilgængelighed
    1 * 1 Items
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • SPME inlet guide for manual injection
    SPME inlet guide for manual injection
    VARISU57356U