Solid phase extraction cartridges, Bond Elut™ C18 OH
Lige cylinderpatron, C18 OH, 150 Å, 3 ml, 500 mg
Katalog # VARI12102046
Leverandør: VARIAN
Gå til:
Product Details & Documents
Bond Elut™ C18 OH is a non-end capped version of the octadecyl bonded phases that enables the silanols on the silica surface to be more active. This low-load C18 has well-controlled silanol activity that permits the fractionation of metabolites and enhances retention of basic compounds compared to an endcapped C18. The pore size makes it a good choice for peptide extraction.
- Silanol activity permits metabolite fractionation
- Tight QC tolerances deliver batch-to-batch reproducibility
- 150 Å pore size expands retention window to higher molecular weight compounds
Specifikationer
- Porestørrelse:150 Å
- Kapacitet:3 ml
- Beskrivelse:Lige cylinderpatron
- Seq:04
- Lejevægt:500 mg
- Fase:C18 OH
- Cat. No.:VARI12102046
- Endcapped:Ingen
- Kulstofbelastning:14,9%
- Surface area:300 m²/g
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



