Particle traps for use with PLOT columns
Particle trap connectors for PoraPLOT columns, UltiMetal, 0,25 mm ID
Catalog # VARICP4795
Supplier: VARIAN
Jump to:
Product Details & Documents
Though highly stabilised, it is impossible to guarantee that no particles will dislodge from the column wall. When used in valve-switching applications, the use of a particle trap can prevent scarring of the rotors.
Specifications
- Beskrivelse:Particle trap connectors for PoraPLOT columns, UltiMetal, 0,25 mm ID
- Seq:08