Orders LinkContactUsLinkComponent
HPLC columns, MonoChrom® C18
Forkolonne, C18, 110 Å, 3 µm
  VARIA0400MG2
 :   VARIAN
HPLC columns, MonoChrom® C18
  VARIA0400MG2
 :  VARIAN
HPLC columns, MonoChrom® C18
Forkolonne, C18, 110 Å, 3 µm
4.400,00 kr
1 * 3 Items
MonoChrom® columns are ideal for separation of organic amines, organic acids, amphoteric compounds and neutral compounds. Advanced silica production and silane bonding techniques allow for the production of material with exceptional base-deactivated characteristics.
  • Kolonne form:
    Forkolonne
  • Partikelstørrelse:
    3 µm
  • Porestørrelse:
    110 Å
  • Seq:
    12
  • Fase:
    C18