Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Bond Elut™ SPEC
Solid phase extraction cartridges, Bond Elut™ SPEC
Disk, C8
Katalog # VARIA74702
Leverandør:   VARIAN
Solid phase extraction cartridges, Bond Elut™ SPEC
Katalog # VARIA74702
Leverandør:  VARIAN
Solid phase extraction cartridges, Bond Elut™ SPEC
Solid phase extraction cartridges, Bond Elut™ SPEC
Disk, C8
Price
2.090,00 kr
Log ind for tilgængelighed
1 * 24 Items
Product Details & Documents
SPEC SPE solid phase extraction cartridges available in standard straight barrel tube format, offering flexibility in sample size. Use on any standard vacuum manifold such as the Vac Elut 20 or SPS 24.
Specifikationer
  • Beskrivelse:
    Disk
  • Seq:
    26
  • Fase:
    C8
Accessories
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Environmental disk manifold
    Environmental disk manifold
    VARIA712
    Log ind for tilgængelighed
    1 * 1 Items - 11.590,00 kr
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Environmental disk holder, 47 mm
    Environmental disk holder, 47 mm
    VARIA713
    Log ind for tilgængelighed
    1 * 1 Items
Produktinformation
  • Katalog #
    Tilgængelighed
    Price
  • Flask, 1 l, male 40/35 mm, ground glass fitting
    Flask, 1 l, male 40/35 mm, ground glass fitting
    VARIA714
    Log ind for tilgængelighed
    1 * 1 Items - 3.880,00 kr