Oligo(dT)₁₈ primer
Oligo(dT)₁₈ primer, 60 µl/30 µg (100 µM), 1 KIT
Katalog # SO131
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Oligo(dT)₁₈ primer is a synthetic single-stranded 18-mer oligonucleotide with 5'- and 3'-hydroxyl ends. The primer is supplied as a ready to use, 20X concentrated aqueous solution. Oligo(dT)₁₈ primer has been functionally tested in first strand cDNA synthesis.
Specifikationer
Specifikationer
- Pakn:1 KIT
- Beskrivelse:Oligo(dT)₁₈ primer, 60 µl/30 µg (100 µM)