Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Saquinavir Related Compounds, Reference Standard

Saquinavir Related Compounds, Reference Standard

Saquinavir Related Compound A, USP reference standard, Vial, 25 mg
Katalog # 1609831.
Leverandør:   USP
Saquinavir Related Compounds, Reference Standard
Katalog # 1609831.
Leverandør:  USP
CAS nummer:  
Saquinavir Related Compounds, Reference Standard

Saquinavir Related Compound A, USP reference standard, Vial, 25 mg

CAS nummer:  
Price
9.210,00 kr
Log ind for tilgængelighed
1 * 25 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

 : US Pharmacopea Reference Standard

Specifikationer

Specifikationer

  • Pakning:
    Vial
  • Pakn:
    25 mg
  • Standard:
    USP reference standard
  • Beskrivelse:
    Saquinavir Related Compound A
  • Cat. No.:
    1609831.
  • Beskrivelse:
    Saquinavir Related Compound A
  • Synonym(s):
    N-tert-butyl-decahydro-2-[2(R)-hydroxy-4-phenyl-3(S)-[[N-(2-quinolylcarbonyl)-D-asparaginyl]amino]butyl]-(4aS,8aS)-isoquinoline-3(S)-carboxamide

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.