Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
MES, anhydrous ≥99%, high purity suitable for buffer solutions

MES, anhydrous ≥99%, high purity suitable for buffer solutions

HDPE bottle, 500 g
Katalog # E183-500G
Leverandør:   VWR Chemicals
MES, anhydrous ≥99%, high purity suitable for buffer solutions
Katalog # E183-500G
Leverandør:  VWR Chemicals
CAS nummer:  4432-31-9
MES, anhydrous ≥99%, high purity suitable for buffer solutions
MES, anhydrous ≥99%, high purity suitable for buffer solutions

HDPE bottle, 500 g

CAS nummer:  4432-31-9
Price
6.490,00 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents
Synonymer:  2-(N-Morpholino)ethanesulphonic acid, 2-Morpholinoethanesulphonic acid
  • CAS nummer:4432-31-9
  • Formel:C₆H₁₃NO₄S
  • MDL nummer:MFCD00006181
  • Molecular Weight:195.24 g/mol
  • Merck Index:13,05929
  • Smeltende Pt:316 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:224-632-3
Specifikationer

Specifikationstestresultater

  • Abs.@290 nm (20%, Water) (w/w):
    <0.05
  • Heavy Metals:
    ≤10 ppm
  • pKa (20 °C):
    5.9 – 6.3
  • Purity:
    ≥99%
  • Solubility (10%, Water):
    Pass
  • Water (KF):
    ≤1%

Specifikationer

  • Pakning:
    HDPE bottle
  • Pakn:
    500 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.