Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

MES, anhydrous ≥99%, high purity suitable for buffer solutions

HDPE bottle, 250 g
Katalog # E183-250G
Leverandør:   VWR Chemicals
MES, anhydrous ≥99%, high purity suitable for buffer solutions
Katalog # E183-250G
Leverandør:  VWR Chemicals
CAS nummer:  4432-31-9
MES, anhydrous ≥99%, high purity suitable for buffer solutions
MES, anhydrous ≥99%, high purity suitable for buffer solutions

HDPE bottle, 250 g

CAS nummer:  4432-31-9
Price
3.590,00 kr
Log ind for tilgængelighed
1 * 250 g
Product Details & Documents
Synonymer:  2-(N-Morpholino)ethanesulphonic acid, 2-Morpholinoethanesulphonic acid
  • CAS nummer:4432-31-9
  • Formel:C₆H₁₃NO₄S
  • MDL nummer:MFCD00006181
  • Molecular Weight:195.24 g/mol
  • Merck Index:13,05929
  • Smeltende Pt:316 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:224-632-3
Specifikationer

Specifikationstestresultater

  • Abs.@290 nm (20%, Water) (w/w):
    <0.05
  • Heavy Metals:
    ≤10 ppm
  • pKa (20 °C):
    5.9 – 6.3
  • Purity:
    ≥99%
  • Solubility (10%, Water):
    Pass
  • Water (KF):
    ≤1%

Specifikationer

  • Pakning:
    HDPE bottle
  • Pakn:
    250 g

Specifikationer