Kolber, pæreformede med standardslib
100 ml, 29/32 NS, 125 mm, 64 mm
Katalog # 201-1044
Leverandør: Lenz Laborglas
Gå til:
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Hals med standardslib. Hvidt mærkningsfelt.
Specifikationer
- Udv.Ø:64 mm
- Kappestr.:29/32 NS
- Kapacitet:100 ml
- Højde:125 mm
- Seq:10