Kolber, pæreformede med standardslib
50 ml, 29/32 NS, 110 mm, 51 mm
Katalog # 201-1043
Leverandør: Lenz Laborglas
Gå til:
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Hals med standardslib. Hvidt mærkningsfelt.
Documents
BSE TSE Glass Columns
BSE/TSE-certifikater
Specifikationer
- Udv.Ø:51 mm
- Kappestr.:29/32 NS
- Kapacitet:50 ml
- Højde:110 mm