Liebig-kondenser med standardslib
250 mm, 19/26 NS, 19/26 NS
Katalog # 201-1170
Leverandør: Lenz Laborglas
Gå til:
Product Details & Documents
DURAN®, borosilicate 3.3 glass, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
For condensing vapours, with fused mantle, cone and socket. Water supplied through plastic connectors with 8 mm external diameter and GL 14 threaded joints.
DIN 12576
Certifications: DIN 12576.
Documents
BSE TSE Glass Columns
BSE/TSE-certifikater
Specifikationer
- Konusstr.:19/26 NS
- Kondenserlængde:250 mm
- Kappestr.:19/26 NS
- Seq:04
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



