Transfer pipettes, fine tip
Fine tip, 3,3 ml, 18 µl, 147 mm, +, Bulk
Catalog # ALPCLW4070
Supplier: Alpha Laboratories
Jump to:
Product Details & Documents
Non toxic, food grade LDPE.
- For careful control and small volume transfers
- Ideal for supernatant removal, slide preparation and gel loading
Specifications
- Pakket:Bulk
- Steril:+
- Kapacitet:3,3 ml
- Beskrivelse:Fine tip
- Længde:147 mm
- Seq:11
- Dråbevol.:18 µl