Supplies for diode array detector (DAD)/ multiple wavelength detector (MWD)
Standard flow cell with RFID tag, 10 mm, 13 μl, 120 bar for G1315C/D, G1365C/D
Katalog # HEWLG1315-60022
Leverandør: AGILENT HEWLETT PACKARD
Supplies for diode array detector (DAD)/ multiple wavelength detector (MWD)
Katalog # HEWLG1315-60022
Leverandør: AGILENT HEWLETT PACKARD
Gå til:
Product Details & Documents
Specifikationer
- Beskrivelse:Standard flow cell with RFID tag, 10 mm, 13 μl, 120 bar for G1315C/D, G1365C/D
- Seq:07