Orders LinkContactUsLinkComponent
Supplies for diode array detector (DAD)/ multiple wavelength detector (MWD)
Standard flow cell with RFID tag, 10 mm, 13 μl, 120 bar for G1315C/D, G1365C/D
Katalog # HEWLG1315-60022
Leverandør:   AGILENT HEWLETT PACKARD
Supplies for diode array detector (DAD)/ multiple wavelength detector (MWD)
Katalog # HEWLG1315-60022
Supplies for diode array detector (DAD)/ multiple wavelength detector (MWD)
Supplies for diode array detector (DAD)/ multiple wavelength detector (MWD)
Standard flow cell with RFID tag, 10 mm, 13 μl, 120 bar for G1315C/D, G1365C/D
Price
16.570,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Beskrivelse:
    Standard flow cell with RFID tag, 10 mm, 13 μl, 120 bar for G1315C/D, G1365C/D
  • Seq:
    07