Orders LinkContactUsLinkComponent
Capillary column ferrules for gas chromatography
Capillary column ferrules for gas chromatography
High temperature PTV inlet, SS/graphite ferrule ID 0,4 mm, column ID 0,53 mm
Katalog # HEWL5188-5314
Leverandør:   AGILENT HEWLETT PACKARD
Capillary column ferrules for gas chromatography
Katalog # HEWL5188-5314
Capillary column ferrules for gas chromatography
Capillary column ferrules for gas chromatography
High temperature PTV inlet, SS/graphite ferrule ID 0,4 mm, column ID 0,53 mm
Price
2.160,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents
Documents
Specifikationer
  • Beskrivelse:
    High temperature PTV inlet, SS/graphite ferrule ID 0,4 mm, column ID 0,53 mm
  • Seq:
    20