Capillaries for LC systems
Flexible capillaries, blue, stainless steel, without fittings, 12,3 μl, 250 mm, I.Ø 0,25 mm
HEWL5065-9979
Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.
Pre-swaged fittings are assembled according to Swagelok specifications.
- Beskrivelse:Flexible capillaries, blue, stainless steel, without fittings, 12,3 μl, 250 mm, I.Ø 0,25 mm
- Seq:94