Powder funnels, PP/PE
253 mm, 50 mm
Katalog # BURK9606-0240
Leverandør: BURKLE
Gå til:
Product Details & Documents
These powder funnels are designed for safe filling of containers.
- PP/PE
9606-0240 with fine sieve made of phosphor-bronze (not for use with foodstuffs).
Specifikationer
- Tragt udv. Ø:253 mm
- Seq:05
- Stilk udv. Ø:50 mm