γ-Linolensyre
Glass bottle for solids, 250 mg
Katalog # CAYM90220-250
Leverandør: Cayman Chemical
Gå til:
Product Details & Documents
Synonymer: cis,cis,cis-6,9,12-Octadecatrienoic acid, γ-Linolenic acid, (6Z,9Z,12Z)-Octadeca-6,9,12-trienoic acid, 6Z,9Z,12Z-Octadecatrienoic acid
- CAS nummer:506-26-3
- Formel:C₁₈H₃₀O₂
- MDL nummer:MFCD00065718
- Molecular Weight:278.44 g/mol
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:250 mg
- Cat. No.:CAYM90220-250



