α-Linolensyre
Glass bottle for solids, 50 mg
CAYM90210-50
: ALA, (9Z,12Z,15Z)-Octadeca-9,12,15-trienoic acid, Linolenic acid, 9Z,12Z,15Z-Octadecatrienoic acid, cis,cis,cis-9,12,15-Octadecatrienoic acid
- :463-40-1
- :C₁₈H₃₀O₂
- :MFCD00065720
- :278.44 g/mol
- :12,05530
- :443 °C (1013 hPa)
- :–9,5 °C
- :0,9141 g/cm³ (18 °C)
- :Stue temperatur
- :207-334-8
- Pakning:Glass bottle for solids
- Pakn:50 mg