α-Linolensyre
Glass bottle for solids, 250 mg
Catalog # CAYM90210-250
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
Synonyms: ALA, (9Z,12Z,15Z)-Octadeca-9,12,15-trienoic acid, Linolenic acid, 9Z,12Z,15Z-Octadecatrienoic acid, cis,cis,cis-9,12,15-Octadecatrienoic acid
- CAS Number:463-40-1
- Formula:C₁₈H₃₀O₂
- MDL Number:MFCD00065720
- Molecular Weight:278.44 g/mol
- Merck Index:12,05530
- Boiling Pt:443 °C (1013 hPa)
- Melting Pt:–9,5 °C
- Density:0,9141 g/cm³ (18 °C)
- Storage Temperature:Stue temperatur
- EINECS:207-334-8
Specifications
Specifications
- Pakning:Glass bottle for solids
- Pakn:250 mg
Specifications
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



