Erucic acid
Glass bottle for solids, 500 mg
Catalog # CAYM90175-500
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
Synonyms: (Z)-Docos-13-enoic acid
- CAS Number:112-86-7
- Formula:C₂₂H₄₂O₂
- MDL Number:MFCD00063188
- Molecular Weight:338.58 g/mol
- EINECS:204-011-3
Specifications
Specifications
- Pakning:Glass bottle for solids
- Pakn:500 mg
- Cat. No.:CAYM90175-500
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



