5,8,11,14-Eicosatetraynoic acid
Glass bottle for solids, 50 mg
Katalog # CAYM90120-50
Leverandør: Cayman Chemical
5,8,11,14-Eicosatetraynoic acid
Katalog # CAYM90120-50
Leverandør: Cayman Chemical
CAS nummer: 1191-85-1
Gå til:
Product Details & Documents
- CAS nummer:1191-85-1
- Formel:C₂₀H₂₄O₂
- MDL nummer:MFCD00036967
- Molecular Weight:296.41 g/mol
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:50 mg
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



