7-Nitroindazol
Glass bottle for solids, 50 mg
Catalog # CAYM81340-50
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
Synonyms: 7-Nitroindazole, 7-NINA, 7-Nitro-1H-indazole
- CAS Number:2942-42-9
- Formula:C₇H₅N₃O₂
- MDL Number:MFCD00022789
- Molecular Weight:163.14 g/mol
- Melting Pt:190 °C
- Density:1,56 g/cm³ (293 °C)
- EINECS:220-934-4
Specifications
Specifications
- Pakning:Glass bottle for solids
- Pakn:50 mg



