S-Isopropylisothiouroniumbromid
Glass bottle for solids, 50 mg
Katalog # CAYM81290-50
Leverandør: Cayman Chemical
S-Isopropylisothiouroniumbromid
Katalog # CAYM81290-50
Leverandør: Cayman Chemical
CAS nummer: 57200-31-4
Gå til:
Product Details & Documents
- CAS nummer:57200-31-4
- Formel:C₄H₁₁BrN₂S
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:50 mg



