S-Isopropylisothiouroniumbromid
Glass bottle for solids, 100 mg
Katalog # CAYM81290-100
Leverandør: Cayman Chemical
S-Isopropylisothiouroniumbromid
Katalog # CAYM81290-100
Leverandør: Cayman Chemical
CAS nummer: 57200-31-4
Gå til:
Product Details & Documents
- CAS nummer:57200-31-4
- Formel:C₄H₁₁BrN₂S
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:100 mg
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



