Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Eddikesyreanhydrid

Eddikesyreanhydrid

500 Tests
Catalog # CAYM400031-5
Flammable
Corrosive
Irritant
Supplier:   Cayman Chemical
Eddikesyreanhydrid
Catalog # CAYM400031-5
Supplier:  Cayman Chemical
CAS Number:  108-24-7
Eddikesyreanhydrid
Eddikesyreanhydrid

500 Tests

CAS Number:  108-24-7
Price
447,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents
Synonyms:  Acetyl oxide, Acetyl acetate
  • CAS Number:108-24-7
  • Formula:(CH₃CO)₂O
  • MDL Number:MFCD00008705
  • Molecular Weight:102.09 g/mol
  • Merck Index:13,00057
  • Boiling Pt:139,9 °C (1013 hPa)
  • Melting Pt:–73 °C
  • Density:1,08 g/cm³ (20 °C)
  • Flash Pt:49 °C
  • Storage Temperature:Stue temperatur
  • EINECS:203-564-8
  • UN:1715
  • ADR:8,II
  • REACH:01-2119486470-36
Specifications

Specifications

  • Pakn:
    500 Tests

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.