Eddikesyreanhydrid
100 Tests
Katalog # CAYM400031-1
Leverandør: Cayman Chemical
Gå til:
Product Details & Documents
Synonymer: Acetyl oxide, Acetyl acetate
- CAS nummer:108-24-7
- Formel:(CH₃CO)₂O
- MDL nummer:MFCD00008705
- Molecular Weight:102.09 g/mol
- Merck Index:13,00057
- Kogende Pt:139,9 °C (1013 hPa)
- Smeltende Pt:–73 °C
- Tæthed:1,08 g/cm³ (20 °C)
- Flash Pt:49 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:203-564-8
- FN:1715
- ADR:8,II
- NÅ:01-2119486470-36
Specifikationer
Specifikationer
- Pakn:100 Tests



