Orders LinkContactUsLinkComponent
Pulverkolber
Pulverkolber
100 ml, 29/32 NS
Catalog # LENZ03043037
Supplier:   Lenz Laborglas
Pulverkolber
Catalog # LENZ03043037
Supplier:  Lenz Laborglas
Pulverkolber
Pulverkolber
100 ml, 29/32 NS
Price
667,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
DURAN®, borosilicate glass, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Especially for rotary evaporators, for drying of powder-like substances. With center neck, conical joint and inscription label.
Documents
Specifications
  • Kappestr.:
    29/32 NS
  • Kapacitet:
    100 ml